Search
Search

Rangap1| Gene Symbol: Rangap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146174.1 | NP_001139646.1 | 1 - 2 | 210 | 140 | |
| GenBank mRNA | AK042930.1 | - | 1 - 2 | 175 | 140 | |
| AK088768.1 | - | 1 - 2 | 132 | 140 | ||
| AK129452.1 | - | 1 - 2 | 139 | 140 | ||
| AK133905.1 | - | 1 - 2 | 175 | 140 | ||
| AK145459.1 | - | 1 - 2 | 131 | 140 | ||
| AK159009.1 | - | 1 - 2 | 163 | 140 | ||
| AK159525.1 | - | 1 - 2 | 175 | 140 | ||
| AK159930.1 | - | 1 - 2 | 166 | 140 | ||
| BC014855.1 | - | 1 - 2 | 166 | 3157225 | 140 | |
| BC052862.1 | - | 1 - 2 | 133 | 30063509 | 140 | |
| BC066213.1 | - | 1 - 2 | 174 | 30466595 | 140 | |
Set Membership: |
|