Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441128_g1
          See other REG3G GE Assays ›
          Gene Symbol:
          Reg3g
          Gene Name:
          regenerating islet-derived 3 gamma
          Gene Aliases:
          AI449515
          Chromosome Location:
          Chr.6: 78466268 - 78468874 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441128_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Reg3g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011260.1 NP_035390.1 5 - 6 484 104
          GenBank mRNA AK008446.1 - 5 - 6 480 104
          AK008608.1 - 5 - 6 480 104
          BC046602.1 - 1 - 2 1801 104
          BC061139.1 - 5 - 6 482 30298640 104
          D63361.1 - 5 - 6 484 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002539.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Reg3g

          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-dependent toll-like receptor signaling pathway
          acute-phase response
          inflammatory response
          positive regulation of keratinocyte proliferation
          negative regulation of keratinocyte differentiation
          defense response to Gram-positive bacterium
          positive regulation of wound healing
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 D63361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 AK008446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 AK008608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 BC046602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 NM_011260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 D63361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 AK008446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 AK008608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 BC046602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P354126 NM_011260.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19625 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178180 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 D63361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 AK008446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 AK008608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 BC046602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 NM_011260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 D63361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 AK008446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 AK008608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 BC046602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P354126 NM_011260.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 D63361.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 AK008446.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 AK008608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 BC046602.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 NM_011260.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 D63361.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 AK008446.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 AK008608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 BC046602.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354126 NM_011260.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24570197 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178180 - -
          Whole Mouse Genome G4122A A_51_P354126 D63361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 AK008446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 AK008608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 BC046602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 NM_011260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 D63361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 AK008446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 AK008608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 BC046602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354126 NM_011260.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116662 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05323946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05323942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05323949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05323938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05323951 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05323950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05323952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05754238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05352530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05541413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05323952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05658878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05323941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05658874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05124478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05754231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05352525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05658877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05658875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05054882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05626512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570162 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545573 - -
          Affymetrix HT MG-430 PM Array Plate 1448872_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545569 - -
          Mouse Genome 430 2.0 Array 1448872_PM_at - -
          Mouse Genome 430 2.0 Array 1448872_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19695_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19695_at - -
          Affymetrix Mouse Clariom S Array PSR0600019604.mm.1 NM_011260.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019604.mm.1 TC0600002539.mm.1 MTA TC
          Mouse Expression Set 430 1448872_PM_at - -
          Mouse Expression Set 430 1448872_at - -
          Murine Genome U74v2 Set 96064_PM_at - -
          Murine Genome U74v2 Set 162187_PM_f_at - -
          Murine Genome U74v2 Set 162187_f_at - -
          Murine Genome U74v2 Set 96064_at - -
          Mouse Genome 430A 2.0 Array 1448872_PM_at - -
          Mouse Genome 430A 2.0 Array 1448872_at - -
          Affymetrix HT Mouse Genome 430A Array 1448872_at - -
          Affymetrix HT Mouse Genome 430A Array 1448872_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 538424 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340520 - -
          Illumina mouse-6 v1.0 expression beadchip 1340520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2662934 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2662934 - -
          mouse-6 v1.1 (Illumina) scl28820.3.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28820.3.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2662934 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2662934 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • 隱私權聲明
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0