Search
Search

Slc7a5| Gene Symbol: Slc7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011404.3 | NP_035534.2 | 3 - 4 | 864 | 110 | |
| GenBank mRNA | AB017189.1 | - | 3 - 4 | 810 | 110 | |
| AB023409.1 | - | 3 - 4 | 792 | 110 | ||
| AK142288.1 | - | 3 - 4 | 865 | 110 | ||
| AK148134.1 | - | 3 - 4 | 534 | 110 | ||
| BC026131.1 | - | 3 - 4 | 873 | 4188854 | 110 | |
Set Membership: |
|