Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441661_g1
          See other TAGLN GE Assays ›
          Gene Symbol:
          Tagln
          Gene Name:
          transgelin
          Gene Aliases:
          Sm22, Sm22a, Ws310
          Chromosome Location:
          Chr.9: 45929628 - 45936058 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441661_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tagln
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011526.5 NP_035656.1 2 - 3 422 64
          GenBank mRNA AK002880.1 - 2 - 3 261 64
          AK077235.1 - 2 - 3 263 64
          BC003795.1 - 2 - 3 251 3600413 64
          L41154.1 - 2 - 3 261 64
          U36588.1 - 2 - 3 261 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002212.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tagln

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          epithelial cell differentiation
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 U36588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 AK077235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 BC003795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 L41154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 NM_011526.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 U36588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 AK077235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 BC003795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 L41154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450527 NM_011526.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163742 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118526 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111954 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 U36588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 AK077235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 BC003795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 L41154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 NM_011526.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 U36588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 AK077235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 BC003795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 L41154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450527 NM_011526.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26476871 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01921409 - -
          Whole Mouse Genome G4122A A_52_P517683 U36588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517683 AK002880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517683 AK077235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517683 BC003795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517683 L41154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 U36588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 AK077235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 BC003795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 L41154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 NM_011526.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 U36588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 AK077235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 BC003795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 L41154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450527 NM_011526.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07504791 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08074850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08074853 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08074857 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07504790 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08074849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26476861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07856280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08166322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07661067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07714447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08092716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07955024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07714449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08166305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07461527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26476843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593128 - -
          Affymetrix HT MG-430 PM Array Plate 1423505_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593123 - -
          Mouse Genome 430 2.0 Array 1423505_PM_at - -
          Mouse Genome 430 2.0 Array 1423505_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21345_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21345_at - -
          Affymetrix Murine Genome 6500A Array Msa16790_at - -
          Affymetrix Mouse Clariom S Array PSR0900018450.mm.1 NM_011526.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018450.mm.1 NM_011526.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018450.mm.1 TC0900002212.mm.1 MTA TC
          Mouse Expression Set 430 1423505_at - -
          Mouse Expression Set 430 1423505_PM_at - -
          Murine Genome U74v2 Set 93541_PM_at - -
          Murine Genome U74v2 Set 93541_at - -
          Mouse Genome 430A 2.0 Array 1423505_PM_at - -
          Mouse Genome 430A 2.0 Array 1423505_at - -
          Affymetrix HT Mouse Genome 430A Array 1423505_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423505_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101240408 - -
          Illumina mouse-6 v1.0 expression beadchip 101240408 - -
          Illumina mouse-6 v1.0 expression beadchip 6660280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243652 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215145 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243652 - -
          mouse-6 v1.1 (Illumina) scl37005.1_98-S - -
          mouse-6 v1.1 (Illumina) scl35919.5_337-S - -
          Illumina Sentrix mouse-6.v1 bead array 101240408 - -
          Illumina Sentrix mouse-6.v1 bead array 6660280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35919.5_337-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101240408 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243652 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215145 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243652 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline