Search
Search

Timm44| Gene Symbol: Timm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011592.2 | NP_035722.2 | 4 - 5 | 410 | 94 | |
| GenBank mRNA | AK153866.1 | - | 4 - 5 | 414 | 94 | |
| BC023454.1 | - | 4 - 5 | 386 | 94 | ||
| BC043479.1 | - | 4 - 5 | 390 | 94 | ||
| BC110677.1 | - | 4 - 5 | 408 | 4979986 | 94 | |
| BC117523.1 | - | 4 - 5 | 400 | 40105563 | 94 | |
| BC117524.1 | - | 4 - 5 | 400 | 40105561 | 94 | |
| CA493459.1 | - | 4 - 5 | 448 | 94 | ||
| U69898.1 | - | 4 - 5 | 400 | 94 | ||
Set Membership: |
|