Search
Search

Tnfrsf1a| Gene Symbol: Tnfrsf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011609.4 | NP_035739.2 | 6 - 7 | 932 | 78 | |
| GenBank mRNA | AK154388.1 | - | 6 - 7 | 902 | 78 | |
| AK159989.1 | - | 6 - 7 | 856 | 78 | ||
| AY541589.1 | - | 6 - 7 | 613 | 78 | ||
| AY541590.1 | - | 6 - 7 | 613 | 78 | ||
| BC004599.1 | - | 6 - 7 | 848 | 3585060 | 78 | |
| BC052675.1 | - | 6 - 7 | 867 | 30061894 | 78 | |
| L26349.1 | - | 6 - 7 | 900 | 78 | ||
| M59377.1 | - | 6 - 7 | 726 | 78 | ||
| M60468.1 | - | 6 - 7 | 825 | 78 | ||
| X57796.1 | - | 6 - 7 | 903 | 78 | ||
| X59238.1 | - | 6 - 7 | 825 | 78 | ||
Set Membership: |
|