Search
Search
| Gene Symbol: Cd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011611.2 | NP_035741.2 | 4 - 5 | 474 | 65 | |
| NM_170702.2 | NP_733803.2 | 4 - 5 | 474 | 65 | ||
| NM_170703.2 | NP_733804.1 | 4 - 5 | 474 | 65 | ||
| NM_170704.2 | NP_733805.1 | 4 - 5 | 474 | 65 | ||
| NR_027852.1 | 4 - 5 | 474 | 65 | |||
| GenBank mRNA | AJ401387.1 | - | 4 - 5 | 408 | 65 | |
| AJ401388.1 | - | 4 - 5 | 408 | 65 | ||
| AJ401389.1 | - | 4 - 5 | 408 | 65 | ||
| AJ401390.1 | - | 4 - 5 | 408 | 65 | ||
| AK089861.1 | - | 427 | 65 | |||
| AK150959.1 | - | 4 - 5 | 431 | 65 | ||
| AK151458.1 | - | 4 - 5 | 431 | 65 | ||
| AK152716.1 | - | 4 - 5 | 432 | 65 | ||
| AK152756.1 | - | 4 - 5 | 430 | 65 | ||
| AK152942.1 | - | 4 - 5 | 438 | 65 | ||
| AK156644.1 | - | 4 - 5 | 442 | 65 | ||
| AK161978.1 | - | 4 - 5 | 431 | 65 | ||
| AK162305.1 | - | 4 - 5 | 484 | 65 | ||
| BC029254.1 | - | 4 - 5 | 432 | 4018221 | 65 | |
| BY743050.1 | - | 4 - 5 | 430 | 65 | ||
| CT010343.1 | - | 4 - 5 | 402 | 65 | ||
| M83312.1 | - | 4 - 5 | 410 | 65 | ||
Set Membership: |
|