Search
Search
| Gene Symbol: Tfrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011638.4 | NP_035768.1 | 11 - 12 | 1496 | 107 | |
| GenBank mRNA | AK048228.1 | - | 11 - 12 | 1410 | 107 | |
| AK088961.1 | - | 11 - 12 | 1410 | 107 | ||
| BC054522.1 | - | 11 - 12 | 1381 | 6406097 | 107 | |
| X57349.1 | - | 10 - 11 | 1332 | 107 | ||
Set Membership: |
|