Search
Search

Trp53| Gene Symbol: Trp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127233.1 | NP_001120705.1 | 8 - 9 | 1063 | 123 | |
| NM_011640.3 | NP_035770.2 | 8 - 9 | 1063 | 123 | ||
| GenBank mRNA | AB017815.1 | - | 8 - 9 | 1007 | 123 | |
| AB017816.1 | - | 8 - 9 | 1007 | 123 | ||
| AB020317.1 | - | 8 - 9 | 1007 | 123 | ||
| AB021961.1 | - | 8 - 9 | 1006 | 123 | ||
| AB221571.1 | - | 7 - 8 | 906 | 123 | ||
| AF051368.1 | - | 8 - 9 | 931 | 123 | ||
| AF151353.1 | - | 8 - 9 | 1028 | 123 | ||
| AF161020.1 | - | 7 - 8 | 916 | 123 | ||
| AK147815.1 | - | 8 - 9 | 1020 | 123 | ||
| AY044188.1 | - | 7 - 8 | 906 | 123 | ||
| AY212017.1 | - | 8 - 9 | 1017 | 123 | ||
| AY423848.1 | - | 1 - 2 | 78 | 123 | ||
| BC005448.1 | - | 8 - 9 | 1034 | 3599812 | 123 | |
| EU031806.1 | - | 8 - 9 | 968 | 123 | ||
| KF766124.1 | - | 8 - 9 | 762 | 123 | ||
| M13872.1 | - | 8 - 9 | 1018 | 123 | ||
| M13873.1 | - | 8 - 9 | 974 | 123 | ||
| M13874.1 | - | 8 - 9 | 959 | 123 | ||
| X00741.1 | - | 8 - 9 | 1025 | 123 | ||
| X01237.1 | - | 8 - 9 | 1063 | 123 | ||
Set Membership: |
|