Search
Search

Sema4g| Gene Symbol: Sema4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011976.1 | NP_036106.1 | 12 - 13 | 2014 | 111 | |
| GenBank mRNA | AF134918.1 | - | 12 - 13 | 2014 | 111 | |
| AK197297.1 | - | 3 - 4 | 329 | 111 | ||
| BC049183.1 | - | 12 - 13 | 2007 | 6395010 | 111 | |
| BC054532.1 | - | 11 - 12 | 2248 | 111 | ||
Set Membership: |
|