Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00442687_m1
          See other APOA2 GE Assays ›
          Gene Symbol:
          Apoa2
          Gene Name:
          apolipoprotein A-II
          Gene Aliases:
          Alp-2, Apo-AII, ApoA-II, ApoAII, Apoa-2, Hdl-1
          Chromosome Location:
          Chr.1: 171225011 - 171226379 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00442687_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apoa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013474.2 NP_038502.2 2 - 3 149 106
          GenBank mRNA AK145823.1 - 2 - 3 112 106
          BC031786.1 - 1 - 2 95 4195958 106
          BU609941.1 - 300 106
          BY780589.1 - 2 - 3 147 106
          CB947382.1 - 2 - 3 114 106
          M79361.1 - 2 - 3 114 106
          M79362.1 - 2 - 3 114 106
          S79794.1 - 1 - 2 55 106
          X04119.1 - 1 - 2 81 106
          X62772.1 - 2 - 3 97 106

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apoa2

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine secretion involved in immune response
          protein folding
          fatty acid metabolic process
          phosphatidylcholine biosynthetic process
          transport
          lipid transport
          cholesterol metabolic process
          phospholipid catabolic process
          response to glucose
          positive regulation of cholesterol esterification
          negative regulation of very-low-density lipoprotein particle remodeling
          beta-glucoside transport
          protein oxidation
          peptidyl-methionine modification
          regulation of intestinal cholesterol absorption
          cholesterol transport
          organ regeneration
          regulation of protein stability
          negative regulation of cholesterol transport
          cholesterol efflux
          phospholipid efflux
          triglyceride-rich lipoprotein particle remodeling
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          high-density lipoprotein particle clearance
          lipoprotein metabolic process
          cholesterol homeostasis
          positive regulation of catalytic activity
          reverse cholesterol transport
          positive regulation of interleukin-8 biosynthetic process
          diacylglycerol catabolic process
          negative regulation of lipid catabolic process
          positive regulation of lipid catabolic process
          negative regulation of lipase activity
          negative regulation of cholesterol import
          negative regulation of cholesterol transporter activity
          lipid transporter activity
          phospholipid binding
          high-density lipoprotein particle binding
          lipid binding
          cholesterol binding
          cholesterol transporter activity
          phosphatidylcholine binding
          apolipoprotein receptor binding
          protein homodimerization activity
          protein heterodimerization activity
          lipase inhibitor activity
          phosphatidylcholine-sterol O-acyltransferase activator activity
          high-density lipoprotein particle receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178553 AK145823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178553 BC031786.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178553 NM_013474.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178553 CB947382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178553 X62772.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966035 AK145823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966035 NM_013474.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966035 CB947382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966035 M79361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966035 X62772.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64590 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174692 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178553 AK145823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178553 BC031786.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178553 NM_013474.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178553 CB947382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178553 X62772.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966035 AK145823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966035 NM_013474.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966035 CB947382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966035 M79361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966035 X62772.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P128973 BC031786.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P128973 M79362.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P128973 S79794.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P128973 X04119.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826747 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174692 - -
          Whole Mouse Genome G4122A A_51_P128973 BC031786.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P128973 M79362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P128973 S79794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P128973 X04119.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116139 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00998718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00998721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00914434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00998736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01055832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351549 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351547 - -
          Affymetrix HT MG-430 PM Array Plate 1417950_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351546 - -
          Mouse Genome 430 2.0 Array 1417950_a_at - -
          Mouse Genome 430 2.0 Array 1417950_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11807_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11807_at - -
          Mouse Expression Set 430 1417950_a_at - -
          Mouse Expression Set 430 1417950_PM_a_at - -
          Murine Genome U74v2 Set 99648_PM_at - -
          Murine Genome U74v2 Set 99648_at - -
          Murine Genome U74v2 Set 161924_f_at - -
          Murine Genome U74v2 Set 161924_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1417950_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417950_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417950_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417950_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6860411 - -
          MouseWG-6 v2 Expression BeadChip 6510364 - -
          Illumina mouse-6 v1.0 expression beadchip 6510364 - -
          Illumina mouse-6 v1.0 expression beadchip 6860411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510364 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775957 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247156 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247156 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775957 - -
          mouse-6 v1.1 (Illumina) scl011807.2_4-S - -
          mouse-6 v1.1 (Illumina) scl000890.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510364 - -
          Illumina Sentrix mouse-6.v1 bead array 6860411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000890.1_7-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl011807.2_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860411 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247156 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775957 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247156 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline