Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00442837_m1
          See other GZMB GE Assays ›
          Gene Symbol:
          Gzmb
          Gene Name:
          granzyme B
          Gene Aliases:
          AI553453, CCP-1/C11, CCP1, Ctla-1, Ctla1, GZB
          Chromosome Location:
          Chr.14: 56258856 - 56262260 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00442837_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gzmb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013542.2 NP_038570.1 4 - 5 708 82
          GenBank mRNA AK157856.1 - 4 - 5 690 82
          AK167770.1 - 4 - 5 671 82
          BC002085.1 - 4 - 5 705 3592898 82
          CT010272.1 - 4 - 5 597 82
          M12302.1 - 4 - 5 612 82
          X04072.1 - 4 - 5 708 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002120.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gzmb

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell mediated cytotoxicity
          proteolysis
          apoptotic process
          granzyme-mediated apoptotic signaling pathway
          protein processing
          cytolysis
          serine-type endopeptidase activity
          protein binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 X04072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 AK157856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 BC002085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 M12302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 NM_013542.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 X04072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 AK157856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 BC002085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 M12302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333274 NM_013542.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264401 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125170 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 X04072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 AK157856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 BC002085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 M12302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 NM_013542.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 X04072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 AK157856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 BC002085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 M12302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333274 NM_013542.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 X04072.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 AK157856.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 BC002085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 M12302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 NM_013542.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 X04072.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 AK157856.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 BC002085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 M12302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P333274 NM_013542.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29622012 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125170 - -
          Whole Mouse Genome G4122A A_51_P333274 X04072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 AK157856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 BC002085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 M12302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 NM_013542.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 X04072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 AK157856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 BC002085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 M12302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333274 NM_013542.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137071 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125170 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11835083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11835064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11835073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11835067 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11835054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29622043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11835094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11835095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11835072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11835055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11835061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11835082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11414215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29622029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621985 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420311 - -
          Affymetrix HT MG-430 PM Array Plate 1419060_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420308 - -
          Mouse Genome 430 2.0 Array 1419060_PM_at - -
          Mouse Genome 430 2.0 Array 1419060_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14939_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14939_at - -
          Affymetrix Mouse Clariom S Array PSR1400016345.mm.1 NM_013542.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016345.mm.1 TC1400002120.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016344.mm.1 NM_013542.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016344.mm.1 TC1400002120.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016342.mm.1 NM_013542.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016342.mm.1 TC1400002120.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016343.mm.1 NM_013542.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016343.mm.1 TC1400002120.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016346.mm.1 NM_013542.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016346.mm.1 TC1400002120.mm.1 MTA TC
          Mouse Expression Set 430 1419060_PM_at - -
          Mouse Expression Set 430 1419060_at - -
          Murine Genome U74v2 Set 102877_at - -
          Murine Genome U74v2 Set 102877_PM_at - -
          Mouse Genome 430A 2.0 Array 1419060_at - -
          Mouse Genome 430A 2.0 Array 1419060_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419060_at - -
          Affymetrix HT Mouse Genome 430A Array 1419060_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 884118 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 380066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000380066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2653555 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2653555 - -
          mouse-6 v1.1 (Illumina) scl45509.5.1_248-S - -
          Illumina Sentrix mouse-6.v1 bead array 380066 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45509.5.1_248-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 380066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2653555 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2653555 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline