Search
Search
| Gene Symbol: Rora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289916.1 | NP_001276845.1 | 2 - 3 | 296 | 76 | |
| NM_013646.2 | NP_038674.1 | 3 - 4 | 305 | 76 | ||
| GenBank mRNA | AK149294.1 | - | 2 - 3 | 186 | 76 | |
| AK155827.1 | - | 2 - 3 | 247 | 76 | ||
| BC003757.1 | - | 2 - 3 | 245 | 3592667 | 76 | |
| S82720.1 | - | 2 - 3 | 222 | 76 | ||
| U53228.1 | - | 3 - 4 | 289 | 76 | ||
| Y08640.1 | - | 2 - 3 | 454 | 76 | ||
Set Membership: |
|