Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00443196_m1
          See other TCF4 GE Assays ›
          Gene Symbol:
          Tcf4
          Gene Name:
          transcription factor 4
          Gene Aliases:
          5730422P05Rik, ASP-I2, E2-2, E2.2, ITF-2, ITF-2b, ITF2, ME2, MITF-2A, MITF-2B, SEF-2, SEF2, SEF2-1, TFE, Tcf-4, bHLHb19
          Chromosome Location:
          Chr.18: 69344490 - 69687967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00443196_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083967.1 NP_001077436.1 2 - 3 310 96
          NM_013685.2 NP_038713.1 2 - 3 625 96
          GenBank mRNA AK028748.1 - 2 - 3 236 96
          AK081012.1 - 2 - 3 262 96
          AK133885.1 - 2 - 3 971 96
          AK145285.1 - 2 - 3 258 96
          BC043050.1 - 1 - 2 85 5708370 96
          S75870.1 - 2 - 3 197 96
          U16322.1 - 2 - 3 597 96
          X91753.1 - 2 - 3 249 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000741.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK145285.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcf4

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          DNA-templated transcription, initiation
          regulation of transcription, DNA-templated
          transcription initiation from RNA polymerase II promoter
          multicellular organism development
          nervous system development
          cell differentiation
          positive regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          protein-DNA complex assembly
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, TFIIB-class binding
          TFIIB-class transcription factor binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          protein C-terminus binding
          protein homodimerization activity
          bHLH transcription factor binding
          protein heterodimerization activity
          protein dimerization activity
          E-box binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800005379.mm.1 NM_001083967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005379.mm.1 NM_001083967.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005379.mm.1 NM_013685.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005379.mm.1 NM_013685.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005379.mm.1 TC1800000741.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005372.mm.1 NM_013685.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005372.mm.1 NM_013685.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005372.mm.1 TC1800000741.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005374.mm.1 NM_001083967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005374.mm.1 NM_001083967.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005374.mm.1 TC1800000741.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005378.mm.1 NM_001083967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005378.mm.1 NM_001083967.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005378.mm.1 NM_013685.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005378.mm.1 NM_013685.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005378.mm.1 TC1800000741.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005373.mm.1 NM_001083967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005373.mm.1 NM_001083967.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005373.mm.1 TC1800000741.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline