Search
Search

Nr2e3| Gene Symbol: Nr2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013708.4 | NP_038736.1 | 6 - 7 | 1142 | 58 | |
| GenBank mRNA | AF148129.1 | - | 6 - 7 | 1004 | 58 | |
| AF204053.1 | - | 6 - 7 | 1142 | 58 | ||
| AK044718.1 | - | 5 - 6 | 1274 | 58 | ||
| AK045788.1 | - | 4 - 5 | 885 | 58 | ||
| AK045859.1 | - | 4 - 5 | 885 | 58 | ||
| AK053560.1 | - | 6 - 7 | 1110 | 58 | ||
| BC017521.1 | - | 6 - 7 | 1136 | 4512088 | 58 | |
| BC047150.1 | - | 3 - 4 | 1829 | 58 | ||
Set Membership: |
|