Search
Search
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016811.2 | NP_058091.2 | 9 - 10 | 1043 | 70 | |
| GenBank mRNA | AF085219.1 | - | 8 - 9 | 729 | 70 | |
| BC006713.1 | - | 9 - 10 | 1043 | 3584592 | 70 | |
| JN832678.1 | - | 9 - 10 | 1062 | 70 | ||
| JN832679.1 | - | 9 - 10 | 1062 | 70 | ||
Set Membership: |
|