Search
Search

Stk39| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016866.2 | NP_058562.1 | 2 - 3 | 464 | 87 | |
| GenBank mRNA | AF099988.1 | - | 2 - 3 | 440 | 87 | |
| AK017292.1 | - | 2 - 3 | 422 | 87 | ||
| AK148456.1 | - | 2 - 3 | 501 | 87 | ||
| BC051964.1 | - | 2 - 3 | 751 | 5698326 | 87 | |
| BC064443.1 | - | 2 - 3 | 464 | 6843981 | 87 | |
Set Membership: |
|