Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444851_m1
          See other TUBD1 GE Assays ›
          Gene Symbol:
          Tubd1
          Gene Name:
          tubulin, delta 1
          Gene Aliases:
          4930550G19Rik, Tubd
          Chromosome Location:
          Chr.11: 86544991 - 86567422 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444851_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tubd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199045.1 NP_001185974.1 4 - 5 927 108
          NM_001199046.1 NP_001185975.1 4 - 5 821 108
          NM_001199047.1 NP_001185976.1 4 - 5 927 108
          NM_019756.3 NP_062730.1 4 - 5 821 108
          GenBank mRNA AB024329.1 - 3 - 4 595 108
          AB031043.1 - 3 - 4 533 108
          AF081568.1 - 4 - 5 661 108
          AK016086.1 - 4 - 5 621 108
          AK030214.1 - 4 - 5 927 108
          AK169454.1 - 4 - 5 822 108
          AK170304.1 - 4 - 5 786 108
          AK171546.1 - 4 - 5 788 108
          BC026465.1 - 4 - 5 804 4484099 108
          BC132325.1 - 4 - 5 654 40130602 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001317.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tubd1

          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          multicellular organism development
          spermatogenesis
          cell differentiation
          protein polymerization
          nucleotide binding
          GTPase activity
          structural constituent of cytoskeleton
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27989558 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098578 BC026465.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098578 AK170304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098578 AK171546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098578 AK216514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098578 AK030214.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303484 AK076507.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303484 NM_007410.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13649 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107528 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098578 BC026465.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098578 AK170304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098578 AK171546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098578 AK216514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098578 AK030214.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302056 BC026465.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302056 BC132325.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302056 AK169454.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302056 AK170304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302056 AK171546.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27989573 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02385379 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07428895 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07428930 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107528 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07428915 - -
          Whole Mouse Genome G4122A A_51_P302056 BC026465.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302056 BC132325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302056 AK169454.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302056 AK170304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302056 AK171546.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136779 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07428915 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107528 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09190959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09665915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09665920 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27989558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27989645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09334018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09553888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09730209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09665915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09266145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09003760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09531899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09414730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09003761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09293246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09190959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27989564 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379973 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379972 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379980 - -
          Affymetrix HT MG-430 PM Array Plate 1421748_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379968 - -
          Mouse Genome 430 2.0 Array 1421748_a_at - -
          Mouse Genome 430 2.0 Array 1421748_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56427_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56427_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_001199045.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_001199045.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_001199046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_001199046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_001199047.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_001199047.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_019756.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 NM_019756.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012345.mm.1 TC1100001317.mm.1 MTA TC
          Mouse Expression Set 430 1421748_a_at - -
          Mouse Expression Set 430 1421748_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1421748_a_at - -
          Mouse Genome 430A 2.0 Array 1421748_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421748_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421748_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 451721 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5340093 - -
          Illumina mouse-6 v1.0 expression beadchip 5340093 - -
          Illumina mouse-6 v1.0 expression beadchip 6290059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2506229 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2464061 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2506229 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2464061 - -
          mouse-6 v1.1 (Illumina) scl41095.10.1_0-S - -
          mouse-6 v1.1 (Illumina) scl0001490.1_34-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290059 - -
          Illumina Sentrix mouse-6.v1 bead array 5340093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001490.1_34-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41095.10.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340093 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2464061 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2506229 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2506229 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2464061 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline