Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445136_s1
          See other P2RY4 GE Assays ›
          Gene Symbol:
          P2ry4
          Gene Name:
          pyrimidinergic receptor P2Y, G-protein coupled, 4
          Gene Aliases:
          P2Y4, P2Y4R
          Chromosome Location:
          Chr.X: 100588358 - 100594927 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445136_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2ry4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020621.4 NP_065646.1 3 349 94
          GenBank mRNA AK076364.1 - 3 350 94
          BC119206.1 - 2 131 8733955 94
          BC120511.1 - 2 131 40129434 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002704.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2ry4

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          transepithelial chloride transport
          cellular response to prostaglandin E stimulus
          signal transducer activity
          G-protein coupled receptor activity
          ATP binding
          pyrimidine nucleotide binding
          G-protein coupled purinergic nucleotide receptor activity
          UTP-activated nucleotide receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407035 NM_020621.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407035 AK076364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407035 NM_020621.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407035 AK076364.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390966 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128191 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407035 NM_020621.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407035 AK076364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407035 NM_020621.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407035 AK076364.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407035 NM_020621.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407035 AK076364.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407035 NM_020621.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P407035 AK076364.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32560394 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128191 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09373917 - -
          Whole Mouse Genome G4122A A_52_P407035 NM_020621.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407035 AK076364.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407035 NM_020621.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407035 AK076364.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138328 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14951627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15111251 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15020357 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15111263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32560308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15152259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15111267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15111241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15038099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15111252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15111265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15111251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15033876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14600194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14599773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32560296 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605941 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605940 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605939 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605942 - -
          Affymetrix HT MG-430 PM Array Plate 1422276_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605938 - -
          Mouse Genome 430 2.0 Array 1422276_PM_at - -
          Mouse Genome 430 2.0 Array 1422276_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57385_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57385_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00016963.mm.1 NM_020621.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016963.mm.1 NM_020621.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016963.mm.1 TC0X00002704.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016961.mm.1 NM_020621.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016961.mm.1 NM_020621.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016961.mm.1 TC0X00002704.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016960.mm.1 NM_020621.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016960.mm.1 NM_020621.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016960.mm.1 TC0X00002704.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016962.mm.1 NM_020621.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016962.mm.1 NM_020621.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016962.mm.1 TC0X00002704.mm.1 MTA TC
          Mouse Expression Set 430 1422276_at - -
          Mouse Expression Set 430 1422276_PM_at - -
          Mouse Genome 430A 2.0 Array 1422276_PM_at - -
          Mouse Genome 430A 2.0 Array 1422276_at - -
          Affymetrix HT Mouse Genome 430A Array 1422276_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422276_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 703711 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 630053 - -
          Illumina mouse-6 v1.0 expression beadchip 630053 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630053 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710340 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710340 - -
          mouse-6 v1.1 (Illumina) scl54001.3.1_196-S - -
          Illumina Sentrix mouse-6.v1 bead array 630053 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54001.3.1_196-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237953 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237953 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline