Search
Search
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022563.2 | NP_072075.2 | 5 - 6 | 739 | 67 | |
| GenBank mRNA | BC138826.1 | - | 6 - 7 | 985 | 8861848 | 67 |
| BC138827.1 | - | 6 - 7 | 880 | 8861849 | 67 | |
| G49238.1 | - | 6 - 7 | 1046 | 67 | ||
| X76505.1 | - | 6 - 7 | 1046 | 67 | ||
Set Membership: |
|