Search
Search

Abcg5| Gene Symbol: Abcg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031884.1 | NP_114090.1 | 3 - 4 | 455 | 80 | |
| GenBank mRNA | AF312713.2 | - | 3 - 4 | 543 | 80 | |
| AK149569.1 | - | 3 - 4 | 426 | 80 | ||
| AY195872.1 | - | 3 - 4 | 543 | 80 | ||
| AY195873.1 | - | 3 - 4 | 543 | 80 | ||
| BC106766.2 | - | 3 - 4 | 432 | 40039484 | 80 | |
Set Membership: |
|