Search
Search

Acox2| Gene Symbol: Acox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161667.1 | NP_001155139.1 | 6 - 7 | 853 | 141 | |
| NM_053115.2 | NP_444345.2 | 6 - 7 | 978 | 141 | ||
| GenBank mRNA | AJ238492.1 | - | 6 - 7 | 895 | 141 | |
| AK144243.1 | - | 6 - 7 | 847 | 141 | ||
| BC021339.1 | - | 6 - 7 | 857 | 5052291 | 141 | |
Set Membership: |
|