Search
Search

Sfxn2| Gene Symbol: Sfxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053196.3 | NP_444426.3 | 10 - 11 | 937 | 107 | |
| GenBank mRNA | AF325261.1 | - | 10 - 11 | 895 | 107 | |
| AK034875.1 | - | 851 | 107 | |||
| AK149277.1 | - | 2769 | 107 | |||
| AK150722.1 | - | 10 - 11 | 888 | 107 | ||
| AK162407.1 | - | 8 - 9 | 2981 | 107 | ||
| BC019808.1 | - | 10 - 11 | 935 | 5043343 | 107 | |
Set Membership: |
|