Search
Search

Sfxn3| Gene Symbol: Sfxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178012.1 | NP_001171483.1 | 5 - 6 | 817 | 85 | |
| NM_001178013.1 | NP_001171484.1 | 6 - 7 | 913 | 85 | ||
| NM_053197.4 | NP_444427.1 | 6 - 7 | 916 | 85 | ||
| GenBank mRNA | AF325262.1 | - | 6 - 7 | 680 | 85 | |
| AK034514.1 | - | 6 - 7 | 633 | 85 | ||
| AK089985.1 | - | 5 - 6 | 578 | 85 | ||
| AK154274.1 | - | 6 - 7 | 686 | 85 | ||
| BC012208.1 | - | 6 - 7 | 818 | 3985155 | 85 | |
Set Membership: |
|