Search
Search

Tm9sf3| Gene Symbol: Tm9sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133352.2 | NP_579930.1 | 3 - 4 | 567 | 108 | |
| GenBank mRNA | AF269151.1 | - | 3 - 4 | 499 | 108 | |
| AK075807.1 | - | 3 - 4 | 535 | 108 | ||
| AK147380.1 | - | 3 - 4 | 562 | 108 | ||
| AK147629.1 | - | 3 - 4 | 561 | 108 | ||
| AK220279.1 | - | 3 - 4 | 543 | 108 | ||
| BC004799.1 | - | 2 - 3 | 196 | 108 | ||
Set Membership: |
|