Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00446819_m1
          See other RIPK2 GE Assays ›
          Gene Symbol:
          Ripk2
          Gene Name:
          receptor (TNFRSF)-interacting serine-threonine kinase 2
          Gene Aliases:
          2210420D18Rik, CARD3, CARDIAK, CCK, D4Bwg0615e, RICK, RIP2
          Chromosome Location:
          Chr.4: 16122741 - 16163679 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00446819_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ripk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138952.3 NP_620402.1 6 - 7 952 96
          GenBank mRNA AF461040.1 - 6 - 7 850 96
          AF487539.1 - 6 - 7 850 96
          AK159593.1 - 6 - 7 976 96
          AK172319.1 - 6 - 7 974 96
          AK172417.1 - 6 - 7 974 96
          BC025611.1 - 4 - 5 470 96
          BC058088.1 - 6 - 7 1097 96
          BC069878.1 - 6 - 7 977 5316066 96

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ripk2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine-mediated signaling pathway
          adaptive immune response
          immune system process
          positive regulation of T-helper 1 type immune response
          protein phosphorylation
          apoptotic process
          I-kappaB kinase/NF-kappaB signaling
          positive regulation of peptidyl-threonine phosphorylation
          phosphorylation
          positive regulation of protein ubiquitination
          lipopolysaccharide-mediated signaling pathway
          positive regulation of chemokine production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of stress-activated MAPK cascade
          positive regulation of immature T cell proliferation
          positive regulation of peptidyl-serine phosphorylation
          toll-like receptor 2 signaling pathway
          toll-like receptor 4 signaling pathway
          T cell proliferation
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to exogenous dsRNA
          innate immune response
          positive regulation of T-helper 1 cell differentiation
          positive regulation of JNK cascade
          positive regulation of alpha-beta T cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing 1 signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          response to interleukin-1
          response to interleukin-12
          response to interleukin-18
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to peptidoglycan
          cellular response to muramyl dipeptide
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          LIM domain binding
          protein homodimerization activity
          CARD domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline