Search
Search

Sptlc1| Gene Symbol: Sptlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009269.2 | NP_033295.2 | 12 - 13 | 1153 | 130 | |
| GenBank mRNA | AF003823.1 | - | 12 - 13 | 1157 | 130 | |
| AK028713.1 | - | 12 - 13 | 1164 | 130 | ||
| AK043064.1 | - | 8 - 9 | 732 | 130 | ||
| AK053207.1 | - | 12 - 13 | 1158 | 130 | ||
| AK079578.1 | - | 12 - 13 | 1154 | 130 | ||
| AK084391.1 | - | 12 - 13 | 1157 | 130 | ||
| AK084445.1 | - | 12 - 13 | 1150 | 130 | ||
| AK089030.1 | - | 12 - 13 | 1157 | 130 | ||
| AK161140.1 | - | 12 - 13 | 1190 | 130 | ||
| AK161811.1 | - | 8 - 9 | 732 | 130 | ||
| BC046323.1 | - | 12 - 13 | 1153 | 6391985 | 130 | |
| BC068243.1 | - | 4011 | 130 | |||
| X95641.1 | - | 12 - 13 | 1152 | 130 | ||
Set Membership: |
|