Search
Search

Pou2f1| Gene Symbol: Pou2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011137.3 | NP_035267.2 | 11 - 12 | 1295 | 64 | |
| NM_198932.3 | NP_945150.2 | 11 - 12 | 1295 | 64 | ||
| NM_198933.3 | NP_945151.2 | 11 - 12 | 1367 | 64 | ||
| NM_198934.3 | NP_945152.2 | 11 - 12 | 1367 | 64 | ||
| GenBank mRNA | AF508939.1 | - | 11 - 12 | 1276 | 64 | |
| AJ489474.1 | - | 11 - 12 | 1367 | 64 | ||
| AJ489475.1 | - | 10 - 11 | 1184 | 64 | ||
| AK028237.1 | - | 11 - 12 | 1297 | 64 | ||
| AY177625.1 | - | 11 - 12 | 1276 | 64 | ||
| BC066201.1 | - | 11 - 12 | 1272 | 64 | ||
| S65461.1 | - | 10 - 11 | 1195 | 64 | ||
| X51958.1 | - | 2 - 3 | 302 | 64 | ||
| X56230.1 | - | 10 - 11 | 1372 | 64 | ||
| X68362.1 | - | 11 - 12 | 1268 | 64 | ||
| X68363.1 | - | 11 - 12 | 1266 | 64 | ||
| X68364.1 | - | 11 - 12 | 1282 | 64 | ||
| X70324.1 | - | 9 - 10 | 1127 | 64 | ||
| X70325.1 | - | 1 - 2 | 108 | 64 | ||
Set Membership: |
|