Search
Search
| Gene Symbol: Prep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011156.2 | NP_035286.1 | 9 - 10 | 1335 | 76 | |
| GenBank mRNA | AB007631.1 | - | 9 - 10 | 1218 | 76 | |
| AK053816.1 | - | 9 - 10 | 1346 | 76 | ||
| AK076257.1 | - | 9 - 10 | 1334 | 76 | ||
| AK133875.1 | - | 9 - 10 | 1334 | 76 | ||
| AK141851.1 | - | 9 - 10 | 1344 | 76 | ||
| AK161528.1 | - | 9 - 10 | 1334 | 76 | ||
| AK163774.1 | - | 1 - 2 | 211 | 76 | ||
| AK163787.1 | - | 9 - 10 | 1347 | 76 | ||
| AK170570.1 | - | 9 - 10 | 1344 | 76 | ||
| AK210695.1 | - | 1 - 2 | 76 | |||
| BC012869.1 | - | 9 - 10 | 1335 | 3967219 | 76 | |
| BC050830.1 | - | 9 - 10 | 1328 | 6519113 | 76 | |
Set Membership: |
|