Search
Search

Ptprr| Gene Symbol: Ptprr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161837.1 | NP_001155309.1 | 3 - 4 | 412 | 75 | |
| NM_011217.2 | NP_035347.1 | 4 - 5 | 1018 | 75 | ||
| GenBank mRNA | AK028054.1 | - | 4 - 5 | 1019 | 75 | |
| BC119230.1 | - | 4 - 5 | 951 | 8733979 | 75 | |
| BC120547.1 | - | 4 - 5 | 951 | 40129470 | 75 | |
| D31898.1 | - | 4 - 5 | 981 | 75 | ||
| Z30313.1 | - | 3 - 4 | 75 | |||
Set Membership: |
|