Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00449026_g1
          See other THBS1 GE Assays ›
          Gene Symbol:
          Thbs1
          Gene Name:
          thrombospondin 1
          Gene Aliases:
          TSP-1, TSP1, Thbs-1, tbsp1
          Chromosome Location:
          Chr.2: 118111890 - 118127133 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00449026_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thbs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011580.3 NP_035710.2 11 - 12 1961 70
          GenBank mRNA AK080686.1 - 11 - 12 1996 70
          AK145202.1 - 11 - 12 1991 70
          AK163092.1 - 11 - 12 1988 70
          BC042422.1 - 11 - 12 1963 4018176 70
          BC050917.1 - 11 - 12 1961 6405710 70
          M87276.1 - 11 - 12 1987 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK089315.1 - Chr. - - [-] on Build 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001729.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thbs1

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          negative regulation of endothelial cell proliferation
          negative regulation of cell-matrix adhesion
          sprouting angiogenesis
          negative regulation of antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
          negative regulation of dendritic cell antigen processing and presentation
          outflow tract morphogenesis
          endocardial cushion development
          growth plate cartilage development
          inflammatory response
          response to unfolded protein
          cell cycle arrest
          cell adhesion
          response to mechanical stimulus
          response to glucose
          positive regulation of endothelial cell migration
          negative regulation of endothelial cell migration
          negative regulation of plasma membrane long-chain fatty acid transport
          negative regulation of nitric oxide mediated signal transduction
          negative regulation of cGMP-mediated signaling
          negative regulation of plasminogen activation
          positive regulation of macrophage chemotaxis
          positive regulation of fibroblast migration
          positive regulation of cell-substrate adhesion
          cell migration
          negative regulation of angiogenesis
          peptide cross-linking
          positive regulation of blood coagulation
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          regulation of cGMP metabolic process
          response to magnesium ion
          negative regulation of interleukin-12 production
          response to endoplasmic reticulum stress
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of phosphorylation
          positive regulation of tumor necrosis factor biosynthetic process
          positive regulation of macrophage activation
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          engulfment of apoptotic cell
          positive regulation of translation
          positive regulation of angiogenesis
          behavioral response to pain
          blood vessel morphogenesis
          positive regulation of chemotaxis
          response to calcium ion
          positive regulation of protein kinase B signaling
          negative regulation of fibrinolysis
          positive regulation of transforming growth factor beta production
          cellular response to nitric oxide
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          negative regulation of endothelial cell chemotaxis
          phosphatidylserine binding
          fibronectin binding
          integrin binding
          calcium ion binding
          protein binding
          heparin binding
          fibroblast growth factor binding
          low-density lipoprotein particle binding
          laminin binding
          transforming growth factor beta binding
          extracellular matrix binding
          fibrinogen binding
          collagen V binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline