Search
Search

Nr2c1| Gene Symbol: Nr2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011629.3 | NP_035759.3 | 7 - 8 | 942 | 72 | |
| GenBank mRNA | AK146891.1 | - | 7 - 8 | 955 | 72 | |
| AK149374.1 | - | 7 - 8 | 857 | 72 | ||
| BC090662.1 | - | 7 - 8 | 918 | 30682075 | 72 | |
| BC094580.1 | - | 7 - 8 | 860 | 30602939 | 72 | |
| L26957.1 | - | 6 - 7 | 751 | 72 | ||
| U28265.1 | - | 6 - 7 | 751 | 72 | ||
| U30482.1 | - | 6 - 7 | 751 | 72 | ||
| Y11436.1 | - | 6 - 7 | 1390 | 72 | ||
Set Membership: |
|