Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00449343_g1
          See other PSMD13 GE Assays ›
          Gene Symbol:
          Psmd13
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, non-ATPase, 13
          Gene Aliases:
          S11
          Chromosome Location:
          Chr.7: 140882394 - 140898642 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00449343_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmd13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011875.4 NP_036005.1 11 - 12 1002 73
          GenBank mRNA AF107838.1 - 11 - 12 963 73
          AK051078.1 - 11 - 12 993 73
          AK075936.1 - 11 - 12 1003 73
          AK081160.1 - 11 - 12 1000 73
          AK145654.1 - 11 - 12 965 73
          AK163017.1 - 11 - 12 997 73
          AK167169.1 - 11 - 12 987 73
          BC029989.1 - 11 - 12 942 73
          BC139184.1 - 11 - 12 980 8862206 73
          BC139187.1 - 11 - 12 980 8862209 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002028.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmd13

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          meiosis I
          endopeptidase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700017808.mm.1 NM_011875.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017808.mm.1 NM_011875.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017808.mm.1 TC0700002028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017814.mm.1 NM_011875.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017814.mm.1 NM_011875.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017814.mm.1 TC0700002028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017811.mm.1 NM_011875.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017811.mm.1 NM_011875.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017811.mm.1 TC0700002028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017805.mm.1 NM_011875.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017805.mm.1 NM_011875.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017805.mm.1 TC0700002028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017810.mm.1 NM_011875.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017810.mm.1 NM_011875.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017810.mm.1 TC0700002028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017809.mm.1 NM_011875.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017809.mm.1 NM_011875.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017809.mm.1 TC0700002028.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline