Search
Search
| Gene Symbol: Sgcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011890.4 | NP_036020.1 | 2 - 3 | 256 | 66 | |
| GenBank mRNA | AB024921.1 | - | 2 - 3 | 270 | 66 | |
| AK014381.2 | - | 2 - 3 | 270 | 66 | ||
| AK132306.1 | - | 2 - 3 | 315 | 66 | ||
| AK169403.1 | - | 2 - 3 | 257 | 66 | ||
| BC052349.1 | - | 2 - 3 | 246 | 6744003 | 66 | |
Set Membership: |
|