Search
Search

Thbs3| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013691.2 | NP_038719.2 | 20 - 21 | 2563 | 64 | |
| GenBank mRNA | BC053023.1 | - | 20 - 21 | 2517 | 6401682 | 64 |
| L04302.1 | - | 20 - 21 | 2543 | 64 | ||
| L24434.1 | - | 20 - 21 | 2540 | 64 | ||
Set Membership: |
|