Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00450226_m1
          See other SCARB1 GE Assays ›
          Gene Symbol:
          Scarb1
          Gene Name:
          scavenger receptor class B, member 1
          Gene Aliases:
          AI120173, CD36, Cd36l1, Cla-1, Cla1, D5Ertd460e, Hdlq1, Hlb398, SR-B1, SR-BI, SRBI, Srb1, mSR-BI
          Chromosome Location:
          Chr.5: 125277087 - 125341111 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00450226_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scarb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205082.1 NP_001192011.1 1 - 2 333 56
          NM_001205083.1 NP_001192012.1 1 - 2 333 56
          NM_016741.2 NP_058021.1 1 - 2 333 56
          GenBank mRNA AK010622.1 - 1 - 2 258 56
          AK028191.1 - 1 - 2 314 56
          AK033114.1 - 1 - 2 258 56
          AK145588.1 - 1 - 2 259 56
          AK154933.1 - 1 - 2 325 56
          AK158122.1 - 1 - 2 258 56
          AK169906.1 - 1 - 2 309 56
          AK171602.1 - 1 - 2 312 56
          BC004656.1 - 1 - 2 313 3498018 56
          CT010222.1 - 1 - 2 121 56
          U37799.1 - 1 - 2 171 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003271.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scarb1

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endothelial cell proliferation
          pattern recognition receptor signaling pathway
          androgen biosynthetic process
          cholesterol catabolic process
          lipid transport
          receptor-mediated endocytosis
          phagocytosis, recognition
          cell adhesion
          positive regulation of triglyceride biosynthetic process
          positive regulation of cholesterol storage
          regulation of phosphatidylcholine catabolic process
          phospholipid transport
          lipopolysaccharide transport
          cholesterol transport
          lipopolysaccharide-mediated signaling pathway
          detection of lipopolysaccharide
          cholesterol efflux
          high-density lipoprotein particle remodeling
          low-density lipoprotein particle clearance
          high-density lipoprotein particle clearance
          vitamin transmembrane transport
          cholesterol homeostasis
          blood vessel endothelial cell migration
          recognition of apoptotic cell
          reverse cholesterol transport
          adhesion of symbiont to host
          regulation of phagocytosis
          positive regulation of nitric-oxide synthase activity
          triglyceride homeostasis
          cholesterol import
          lipopolysaccharide binding
          phosphatidylserine binding
          lipopolysaccharide receptor activity
          high-density lipoprotein particle binding
          low-density lipoprotein particle binding
          apolipoprotein binding
          apolipoprotein A-I binding
          protein homodimerization activity
          high-density lipoprotein particle receptor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500029610.mm.1 NM_001205082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029610.mm.1 NM_001205082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029610.mm.1 NM_001205083.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029610.mm.1 NM_001205083.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029610.mm.1 NM_016741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029610.mm.1 NM_016741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029610.mm.1 TC0500003271.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029609.mm.1 NM_001205082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029609.mm.1 NM_001205082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029609.mm.1 NM_001205083.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029609.mm.1 NM_001205083.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029609.mm.1 NM_016741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029609.mm.1 NM_016741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029609.mm.1 TC0500003271.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029617.mm.1 NM_001205082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029617.mm.1 NM_001205082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029617.mm.1 NM_001205083.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029617.mm.1 NM_001205083.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029617.mm.1 NM_016741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029617.mm.1 NM_016741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029617.mm.1 TC0500003271.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029618.mm.1 NM_001205082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029618.mm.1 NM_001205082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029618.mm.1 NM_001205083.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029618.mm.1 NM_001205083.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029618.mm.1 NM_016741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029618.mm.1 NM_016741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029618.mm.1 TC0500003271.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029607.mm.1 NM_001205082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029607.mm.1 NM_001205082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029607.mm.1 NM_001205083.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029607.mm.1 NM_001205083.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029607.mm.1 NM_016741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029607.mm.1 NM_016741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029607.mm.1 TC0500003271.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 856472 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline