Search
Search
| Gene Symbol: Stx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018768.2 | NP_061238.1 | 5 - 6 | 473 | 99 | |
| GenBank mRNA | AB040054.1 | - | 5 - 6 | 470 | 99 | |
| AF036716.1 | - | 5 - 6 | 460 | 99 | ||
| AF093064.1 | - | 5 - 6 | 482 | 99 | ||
| AK003297.1 | - | 5 - 6 | 499 | 99 | ||
| AK018716.1 | - | 4 - 5 | 300 | 99 | ||
| BC038186.1 | - | 5 - 6 | 472 | 99 | ||
| BC048167.1 | - | 5 - 6 | 473 | 6821351 | 99 | |
| BC048479.1 | - | 5 - 6 | 498 | 6774827 | 99 | |
Set Membership: |
|