Search
Search
| Gene Symbol: Stx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021433.3 | NP_067408.1 | 3 - 4 | 491 | 81 | |
| GenBank mRNA | AF237814.1 | - | 3 - 4 | 301 | 81 | |
| AK009690.1 | - | 3 - 4 | 446 | 81 | ||
| AK018167.1 | - | 3 - 4 | 479 | 81 | ||
| AK019106.1 | - | 3 - 4 | 453 | 81 | ||
| AK031673.1 | - | 3 - 4 | 470 | 81 | ||
| AK044905.1 | - | 3 - 4 | 452 | 81 | ||
| AK049124.1 | - | 3 - 4 | 451 | 81 | ||
| AK151549.1 | - | 3 - 4 | 445 | 81 | ||
| AK159392.1 | - | 3 - 4 | 463 | 81 | ||
| BC029205.1 | - | 3 - 4 | 408 | 4039560 | 81 | |
| BQ749205.1 | - | 3 - 4 | 464 | 81 | ||
Set Membership: |
|