Search
Search

Tmem8| Gene Symbol: Tmem8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021793.2 | NP_068565.1 | 6 - 7 | 1351 | 65 | |
| GenBank mRNA | AB045293.1 | - | 6 - 7 | 1350 | 65 | |
| AK044384.1 | - | 6 - 7 | 1352 | 65 | ||
| BC005722.1 | - | 5 - 6 | 1227 | 3595205 | 65 | |
Set Membership: |
|