Search
Search

Slc13a2| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022411.3 | NP_071856.1 | 9 - 10 | 1311 | 83 | |
| GenBank mRNA | AF201903.1 | - | 9 - 10 | 1311 | 83 | |
| AK140972.1 | - | 9 - 10 | 1362 | 83 | ||
| BC013493.1 | - | 9 - 10 | 1336 | 4218337 | 83 | |
Set Membership: |
|