Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452101_m1
          See other NGB GE Assays ›
          Gene Symbol:
          Ngb
          Gene Name:
          neuroglobin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 87097531 - 87102539 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452101_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ngb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001294308.1 NP_001281237.1 1 - 2 364 83
          NM_022414.2 NP_071859.1 1 - 2 364 83
          GenBank mRNA AJ245945.1 - 1 - 2 161 83
          AK140173.1 - 1 - 2 348 83
          AK165775.1 - 1 - 2 170 83
          BC024263.1 - 1 - 2 364 5133442 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002114.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ngb

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          apoptotic process
          visual perception
          oxygen transport
          neuron projection development
          positive regulation of catalytic activity
          negative regulation of hydrogen peroxide-induced cell death
          oxygen transporter activity
          iron ion binding
          protein binding
          heme binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614147 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070641 AK140173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070641 AK165775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070641 BC024263.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070641 NM_022414.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129564 AK140173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129564 AK165775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129564 BC024263.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129564 NM_022414.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110650 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159775 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129564 AK140173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129564 AK165775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129564 BC024263.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129564 NM_022414.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070641 AK140173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070641 AK165775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070641 BC024263.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070641 NM_022414.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614182 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159775 - -
          Whole Mouse Genome G4122A A_51_P158734 AK140173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158734 AK165775.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158734 BC024263.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158734 AJ245945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158734 NM_022414.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129564 AK140173.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129564 AK165775.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129564 BC024263.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129564 NM_022414.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09805521 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10407306 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614147 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10364382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10247801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10421030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09779174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10313040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10364378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10421029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10270715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09889997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09805523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09779162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09779167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09853888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10313038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09890526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09853889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10270714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09890523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614174 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10401710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401709 - -
          Affymetrix HT MG-430 PM Array Plate 1417996_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417997_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10401708 - -
          Mouse Genome 430 2.0 Array 1417997_PM_at - -
          Mouse Genome 430 2.0 Array 1417997_at - -
          Mouse Genome 430 2.0 Array 1417996_at - -
          Mouse Genome 430 2.0 Array 1417996_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64242_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64242_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200014646.mm.1 NM_022414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014646.mm.1 NM_022414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014646.mm.1 TC1200002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014647.mm.1 NM_022414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014647.mm.1 NM_022414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014647.mm.1 TC1200002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014648.mm.1 NM_022414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014648.mm.1 NM_022414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014648.mm.1 TC1200002114.mm.1 MTA TC
          Mouse Expression Set 430 1417996_at - -
          Mouse Expression Set 430 1417997_at - -
          Mouse Expression Set 430 1417996_PM_at - -
          Mouse Expression Set 430 1417997_PM_at - -
          Mouse Genome 430A 2.0 Array 1417997_PM_at - -
          Mouse Genome 430A 2.0 Array 1417996_at - -
          Mouse Genome 430A 2.0 Array 1417997_at - -
          Mouse Genome 430A 2.0 Array 1417996_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417997_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417996_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417996_at - -
          Affymetrix HT Mouse Genome 430A Array 1417997_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 425519 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4120575 - -
          Illumina mouse-6 v1.0 expression beadchip 4120575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615819 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2615819 - -
          mouse-6 v1.1 (Illumina) scl42210.5_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 4120575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42210.5_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2615821 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2895873 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2615821 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2895873 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline