Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022653.4 | NP_073144.3 | 8 - 9 | 1436 | 63 | |
GenBank mRNA | AF314187.1 | - | 8 - 9 | 1394 | 63 | |
AK028609.1 | - | 8 - 9 | 1370 | 63 | ||
BC031722.1 | - | 8 - 9 | 1402 | 4019169 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |