Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452214_m1
          See other TSC1 GE Assays ›
          Gene Symbol:
          Tsc1
          Gene Name:
          tuberous sclerosis 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 28641190 - 28691172 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452214_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289575.1 NP_001276504.1 9 - 10 1136 70
          NM_001289576.1 NP_001276505.1 9 - 10 1136 70
          NM_022887.4 NP_075025.2 9 - 10 1107 70
          GenBank mRNA AB047561.1 - 7 - 8 906 70
          AJ271912.1 - 9 - 10 1128 70
          AK030318.1 - 9 - 10 1103 70
          AK083363.1 - 9 - 10 1091 70
          AK147428.1 - 9 - 10 1127 70
          AK154416.1 - 9 - 10 1207 70
          BC052399.1 - 8 - 9 1060 5720967 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000490.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsc1

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          neural tube closure
          regulation of cell-matrix adhesion
          rRNA export from nucleus
          regulation of translation
          potassium ion transport
          cell-matrix adhesion
          nervous system development
          negative regulation of cell proliferation
          embryo development
          negative regulation of translation
          hippocampus development
          cerebral cortex development
          cell projection organization
          negative regulation of TOR signaling
          response to insulin
          regulation of actin cytoskeleton organization
          myelination
          regulation of GTPase activity
          regulation of phosphoprotein phosphatase activity
          negative regulation of cell size
          regulation of protein kinase activity
          glucose import
          negative regulation of insulin receptor signaling pathway
          synapse organization
          protein stabilization
          protein heterooligomerization
          regulation of stress fiber assembly
          regulation of cell cycle
          regulation of focal adhesion assembly
          positive regulation of focal adhesion assembly
          cardiac muscle cell differentiation
          activation of GTPase activity
          protein binding
          GTPase regulator activity
          GTPase activating protein binding
          protein N-terminus binding
          chaperone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200003477.mm.1 NM_001289575.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003477.mm.1 NM_001289576.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003477.mm.1 NM_022887.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003477.mm.1 TC0200000490.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200003476.mm.1 NM_001289575.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003476.mm.1 NM_001289576.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003476.mm.1 NM_022887.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003476.mm.1 TC0200000490.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline