Search
Search

Usp15| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301628.1 | NP_001288557.1 | 11 - 12 | 1568 | 85 | |
| NM_027604.4 | NP_081880.2 | 12 - 13 | 1655 | 85 | ||
| GenBank mRNA | AF468037.1 | - | 12 - 13 | 1559 | 85 | |
| AK014891.1 | - | 6 - 7 | 866 | 85 | ||
| AK122301.1 | - | 11 - 12 | 1483 | 85 | ||
| AK133852.1 | - | 11 - 12 | 1477 | 85 | ||
| AK145749.1 | - | 12 - 13 | 1559 | 85 | ||
| AK161469.1 | - | 12 - 13 | 1598 | 85 | ||
| AK161957.1 | - | 11 - 12 | 1516 | 85 | ||
| AK163315.1 | - | 11 - 12 | 1497 | 85 | ||
| AK168755.1 | - | 11 - 12 | 1503 | 85 | ||
| BC050042.1 | - | 12 - 13 | 1571 | 30022307 | 85 | |
Set Membership: |
|