Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00453173_g1
          See other PACSIN3 GE Assays ›
          Gene Symbol:
          Pacsin3
          Gene Name:
          protein kinase C and casein kinase substrate in neurons 3
          Gene Aliases:
          4921507A02Rik, 6330413E15Rik, AW413130
          Chromosome Location:
          Chr.2: 91256098 - 91264680 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00453173_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pacsin3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289677.1 NP_001276606.1 4 - 5 662 65
          NM_001289678.1 NP_001276607.1 5 - 6 661 65
          NM_028733.4 NP_083009.1 5 - 6 791 65
          NM_030880.3 NP_112019.2 5 - 6 657 65
          GenBank mRNA AF149824.1 - 5 - 6 575 65
          AF242531.1 - 4 - 5 560 65
          AK049166.1 - 5 - 6 792 65
          AK141063.1 - 5 - 6 634 65
          BC003884.1 - 5 - 6 598 3497435 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001374.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pacsin3

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          cytoskeleton organization
          negative regulation of endocytosis
          positive regulation of membrane protein ectodomain proteolysis
          negative regulation of calcium ion transport
          membrane tubulation
          protein binding
          phospholipid binding
          cytoskeletal protein binding
          lipid binding
          calcium channel inhibitor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200010629.mm.1 NM_001289677.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010629.mm.1 NM_001289678.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010629.mm.1 NM_028733.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010629.mm.1 NM_030880.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010629.mm.1 TC0200001374.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010632.mm.1 NM_001289677.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010632.mm.1 NM_001289678.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010632.mm.1 NM_028733.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010632.mm.1 NM_030880.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010632.mm.1 TC0200001374.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010628.mm.1 NM_001289677.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010628.mm.1 NM_001289678.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010628.mm.1 NM_028733.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010628.mm.1 NM_030880.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010628.mm.1 TC0200001374.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010631.mm.1 NM_001289677.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010631.mm.1 NM_001289678.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010631.mm.1 NM_028733.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010631.mm.1 NM_030880.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010631.mm.1 TC0200001374.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010630.mm.1 NM_001289677.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010630.mm.1 NM_001289678.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010630.mm.1 NM_028733.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010630.mm.1 NM_030880.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010630.mm.1 TC0200001374.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline