Search
Search

Naa15| Gene Symbol: Naa15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053089.3 | NP_444319.3 | 15 - 16 | 2256 | 99 | |
| GenBank mRNA | AB221683.1 | - | 15 - 16 | 1949 | 99 | |
| AF237622.1 | - | 11 - 12 | 1540 | 99 | ||
| AF510858.1 | - | 15 - 16 | 2146 | 99 | ||
| BC030167.1 | - | 3 - 4 | 464 | 99 | ||
| BC050017.1 | - | 15 - 16 | 2147 | 30020858 | 99 | |
Set Membership: |
|