Search
Search

Pcnxl3| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144868.3 | NP_659117.2 | 33 - 34 | 6355 | 74 | |
| GenBank mRNA | AF237953.1 | - | 31 - 32 | 4372 | 74 | |
| AK144114.1 | - | 9 - 10 | 1615 | 74 | ||
| AK220471.1 | - | 11 - 12 | 2253 | 74 | ||
| BC038056.1 | - | 28 - 29 | 4463 | 74 | ||
| BC054751.1 | - | 33 - 34 | 5634 | 74 | ||
| BC055289.1 | - | 34 - 35 | 6158 | 74 | ||
| BC096538.1 | - | 33 - 34 | 6355 | 74 | ||
Set Membership: |
|