Search
Search
| Gene Symbol: Nek9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145138.2 | NP_660120.2 | 16 - 17 | 2142 | 61 | |
| GenBank mRNA | AJ489828.1 | - | 16 - 17 | 2117 | 61 | |
| AK173323.1 | - | 16 - 17 | 2126 | 61 | ||
| BC117971.2 | - | 16 - 17 | 2083 | 40098375 | 61 | |
| BC117972.2 | - | 16 - 17 | 2082 | 40098378 | 61 | |
Set Membership: |
|