Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455363_g1
          See other DCLRE1C GE Assays ›
          Gene Symbol:
          Dclre1c
          Gene Name:
          DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae)
          Gene Aliases:
          9930121L06Rik, AI661365, Art, Snm1l
          Chromosome Location:
          Chr.2: 3424062 - 3474330 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455363_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dclre1c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110214.1 NP_001103684.1 7 - 8 620 70
          NM_146114.3 NP_666226.2 7 - 8 620 70
          NM_175683.4 NP_783614.1 7 - 8 620 70
          GenBank mRNA AF387731.1 - 7 - 8 602 70
          AK037126.1 - 7 - 8 587 70
          AK038788.1 - 7 - 8 550 70
          AK052369.1 - 7 - 8 576 70
          AK052490.1 - 7 - 8 559 70
          AK088810.1 - 5 - 6 402 70
          BC108935.2 - 7 - 8 573 40039591 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000006.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dclre1c

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          immune system process
          DNA metabolic process
          DNA repair
          double-strand break repair
          DNA recombination
          cellular response to DNA damage stimulus
          response to ionizing radiation
          B cell differentiation
          V(D)J recombination
          chromosome organization
          nucleic acid phosphodiester bond hydrolysis
          single-stranded DNA endodeoxyribonuclease activity
          nuclease activity
          endonuclease activity
          exonuclease activity
          5'-3' exonuclease activity
          hydrolase activity
          phosphoric ester hydrolase activity
          T/G mismatch-specific endonuclease activity
          retroviral 3' processing activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200000073.mm.1 NM_001110214.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000073.mm.1 NM_001110214.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000073.mm.1 NM_146114.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000073.mm.1 NM_146114.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000073.mm.1 NM_175683.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000073.mm.1 NM_175683.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000073.mm.1 TC0200000006.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000074.mm.1 NM_001110214.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000074.mm.1 NM_001110214.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000074.mm.1 NM_146114.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000074.mm.1 NM_146114.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000074.mm.1 NM_175683.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000074.mm.1 NM_175683.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000074.mm.1 TC0200000006.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline