Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455615_s1
          See other OLFR559 GE Assays ›
          Gene Symbol:
          Olfr559 Olfr33
          Gene Name:
          olfactory receptor 559
          olfactory receptor 33
          Gene Aliases:
          MOR11-1
          MOR11-2, MTPCR33, ORL181
          Chromosome Location:
          Chr.7: 102723532 - 102724488 on Build GRCm38
          Chromosome Location:
          Chr.7: 102713455 - 102714411 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00455615_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr559
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147112.1 NP_667323.1 1 635 91
          GenBank mRNA BC119185.1 - 1 657 8733934 91
          BC120640.1 - 1 657 40129563 91
          Gene Symbol: Olfr33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147073.1 NP_667284.1 1 91
          GenBank mRNA BC104110.1 - 1 40043836 91
          BC104111.1 - 1 40043837 91
          X89685.1 - 1 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003867.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr559
          Click here to find siRNAs for Gene Olfr33

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952775 NM_147112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952775 BC119185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952775 BC120640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P189676 NM_147112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P189676 BC119185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P189676 BC120640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P189676 NM_147112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P189676 BC119185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P189676 BC120640.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674009 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137863 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952775 NM_147112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952775 BC119185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952775 BC120640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P189676 NM_147112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P189676 BC119185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P189676 BC120640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P189676 NM_147112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P189676 BC119185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P189676 BC120640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189676 NM_147112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189676 BC119185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189676 BC120640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189676 NM_147112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189676 BC119185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189676 BC120640.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25380351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137863 - -
          Whole Mouse Genome G4122A A_52_P189676 NM_147112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P189676 BC119185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P189676 BC120640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P189676 NM_147112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P189676 BC119185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P189676 BC120640.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118817 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137863 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06146851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06641812 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06641795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06353340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06641813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06641799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380357 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566171 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566170 - -
          Affymetrix Mouse Clariom S Array PSR0700032658.mm.1 TC0700003867.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900559 - -
          ABI Mouse Genome Survey Microarray 901873 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4920594 - -
          Illumina mouse-6 v1.0 expression beadchip 4920594 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227132 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227132 - -
          mouse-6 v1.1 (Illumina) scl8892.1.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8892.1.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2811336 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2811339 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2811339 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2811336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline